missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Chloro-4-fluorophenyl)indole, 98%

Product Code. 11439743
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439743 1 g 1g
11449743 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439743 Supplier Thermo Scientific Alfa Aesar Supplier No. A12920.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1868-88-8 | C14H9ClFN | 245.681 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1868-88-8
Molecular Formula C14H9ClFN
Molecular Weight (g/mol) 245.681
MDL Number MFCD00068179
InChI Key QNYJRNHACDLRSI-UHFFFAOYSA-N
PubChem CID 2736577
IUPAC Name 2-(3-chloro-4-fluorophenyl)-1H-indole
SMILES C1=CC=C2C(=C1)C=C(N2)C3=CC(=C(C=C3)F)Cl

Specifications

Melting Point 170°C to 173°C
Quantity 1 g
Synonym 2-3-chloro-4-fluorophenyl indole, 2-3-chloro-4-fluorophenyl-1h-indole, 2-3-chloro-4-fluoro-phenyl-1h-indole, 1h-indole, 2-3-chloro-4-fluorophenyl, acmc-20aoqa, pubchem7289, maybridge1_007663, 2-3-chloro-4-fluoro-phenyl indole, 1h-indole,2-3-chloro-4-fluorophenyl, 2-3-chloranyl-4-fluoranyl-phenyl-1h-indole
Formula Weight 245.69
Percent Purity 98%
Chemical Name or Material 2-(3-Chloro-4-fluorophenyl)indole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.