missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Chlorophenoxy)ethanethioamide, ≥95%, Thermo Scientific™

Product Code. 10233572
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10233572 1 g Amber Glass Bottle 1g
10243572 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10233572 Supplier Thermo Scientific Maybridge Supplier No. SPB01327DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 35370-95-7
Molecular Formula C8H8ClNOS
Molecular Weight (g/mol) 201.668
MDL Number MFCD00052479
InChI Key RPAOLVIADVQKNA-UHFFFAOYSA-N
Synonym 2-3-chlorophenoxy ethanethioamide, 2-3-chlorophenoxy thioacetamide, 1-amino-2-3-chlorophenoxy ethane-1-thione, maybridge1_008529, 3-chlorophenoxy thioacetamide, 2-3-chlorophenoxy ethanethioamide #, ethanethioamide,2-3-chlorophenoxy
PubChem CID 2743553
IUPAC Name 2-(3-chlorophenoxy)ethanethioamide
SMILES C1=CC(=CC(=C1)Cl)OCC(=S)N

Specifications

CAS Min % 95.0
Assay Percent Range 95%+
Packaging Amber Glass Bottle
Linear Formula ClC6H4OCH2CSNH2
Quantity 1 g
Formula Weight 201.68
Percent Purity 95%
Chemical Name or Material 2-(3-Chlorophenoxy)ethanethioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.