missing translation for 'onlineSavingsMsg'
Learn More

(2-(3-Chlorophenyl)-1,3-thiazol-4-yl)methanamine hydrochloride monohydrate, 97%, Thermo Scientific™

Product Code. 10433183
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10433183 250 mg 250mg
10699514 1 g 1g
10327644 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10433183 Supplier Thermo Scientific Maybridge Supplier No. MO01298CB
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-12-3
Molecular Formula C10H10Cl2N2S
Molecular Weight (g/mol) 261.164
MDL Number MFCD06738410
InChI Key DBEFCUHDXFEPSJ-UHFFFAOYSA-N
Synonym 2-3-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride, 2-3-chlorophenyl thiazol-4-yl methylamine hydrochloride, c-2-3-chloro-phenyl-thiazol-4-yl-methylamine hydrochloride, 2-3-chlorophenyl-1,3-thiazol-4-yl methane amine hydrochloride 1-hydrate, 4-aminomethyl-2-3-chlorophenyl thiazole hcl, 2-3-chlorophenyl thiazol-4-yl methanamine hydrochloride, 2-3-chlorophenyl-4-thiazolyl methanamine hydrochloride, 2-3-chloro-phenyl-thiazol-4-ylmethyllamine hcl, 2-3-chlorophenyl-1,3-thiazol-4-yl methylamine, chloride, 1-2-3-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride
PubChem CID 24229776
IUPAC Name [2-(3-chlorophenyl)-1,3-thiazol-4-yl]methanamine;hydrochloride
SMILES C1=CC(=CC(=C1)Cl)C2=NC(=CS2)CN.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 279.2
Percent Purity 97%
Chemical Name or Material [2-(3-Chlorophenyl)-1, 3-thiazol-4-yl]methanamine hydrochloride monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.