missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Chlorophenyl)-1,3-thiazole-4-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10709405
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10709405 10 g 10g
10691245 250 mg 250mg
10197282 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10709405 Supplier Thermo Scientific Maybridge Supplier No. MO01112EA
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 845885-82-7
Molecular Formula C10H6ClNO2S
Molecular Weight (g/mol) 239.67
MDL Number MFCD05664421
InChI Key DNSCDQVZFMLMAD-UHFFFAOYSA-N
Synonym 2-3-chlorophenyl-1,3-thiazole-4-carboxylic acid, 2-3-chlorophenyl thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-3-chlorophenyl, 2-3-chlorophenyl thiazole-4-carboxylicacid, 2-3-chlorophenyl-4-thiazolecarboxylic acid, 2-3-chlorophenyl-1 pound not3-thiazole-4-carboxylic acid
PubChem CID 2794667
IUPAC Name 2-(3-chlorophenyl)-1,3-thiazole-4-carboxylic acid
SMILES OC(=O)C1=CSC(=N1)C1=CC(Cl)=CC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 239.68
Percent Purity 97%
Chemical Name or Material 2-(3-Chlorophenyl)-1, 3-thiazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.