missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromobutyric acid, 97%

Product Code. 11341727
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11341727

Brand: Thermo Scientific Alfa Aesar L04841.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 600-30-6 | C4H6Br2O2 | 245.898 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 600-30-6
Molecular Formula C4H6Br2O2
Molecular Weight (g/mol) 245.898
MDL Number MFCD00014334
InChI Key HESQKTULJLBDRF-UHFFFAOYSA-N
Synonym 2,3-dibromobutyric acid, .alpha.,.beta.-dibromobutyric acid, 2,3-dibromo-butyric acid, acmc-1atov, 2,3 dibromobutyric acid, #, butanoic acid,2,3-dibromo, butanoic acid, 2,3-dibromo, alpha,beta-dibromo-n-butyric acid, .alpha.,.beta.-dibromo-n-butyric acid
PubChem CID 95386
IUPAC Name 2,3-dibromobutanoic acid
SMILES CC(C(C(=O)O)Br)Br

Specifications

Melting Point 84°C to 88°C
Boiling Point 100°C to 110°C (2 mmHg)
Quantity 25 g
UN Number UN3261
Beilstein 1721669
Formula Weight 245.91
Percent Purity 97%
Chemical Name or Material 2,3-Dibromobutyric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.