missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromofuran, 97%, stab. with 0.5% calcium carbonate

Product Code. 11348306
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348306 5 g 5g
11338306 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348306 Supplier Thermo Scientific Alfa Aesar Supplier No. L15518.06
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 30544-34-4 | C4H2Br2O | 225.867 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30544-34-4
Molecular Formula C4H2Br2O
Molecular Weight (g/mol) 225.867
MDL Number MFCD01074839
InChI Key GKPGEBCMRMQOPF-UHFFFAOYSA-N
Synonym 2,3-dibrom-furan, acmc-1adxx, furan, 2,3-dibromo, 2,3-bis bromanyl furan
PubChem CID 7021501
IUPAC Name 2,3-dibromofuran
SMILES C1=COC(=C1Br)Br

Specifications

Density 2
Boiling Point 75°C to 78°C (75 mmHg)
Refractive Index 1.547
Quantity 5 g
Beilstein 107511
Sensitivity Light sensitive
Formula Weight 225.87
Percent Purity 97%
Chemical Name or Material 2,3-Dibromofuran, stabilized with 0.5% calcium carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.