missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloroaniline, 99%

Product Code. 11404330
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404330 100 g 100g
11414330 500 g 500g
11494320 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404330 Supplier Thermo Scientific Alfa Aesar Supplier No. A11524.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 608-27-5 | C6H5Cl2N | 162.013 g/mol

2,3-Dichloroaniline is used in dyestuffs, pigments and optical brighteners. It is used as flame retardants for polymers. It is used in the manufacturing of growth regulators, polymer auxiliaries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dichloroaniline is used in dyestuffs, pigments and optical brighteners. It is used as flame retardants for polymers. It is used in the manufacturing of growth regulators, polymer auxiliaries.

Solubility
Soluble in ether, alcohol and acetone. Slightly soluble in benzene. Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-27-5
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00007657
InChI Key BRPSAOUFIJSKOT-UHFFFAOYSA-N
Synonym benzenamine, 2,3-dichloro, 2,3-dichlorobenzenamine, aniline, 2,3-dichloro, unii-2bl4f1dxvn, 2,3-dichloranilin, 2,3 dichloraniline, 2,3-dichloro-phenylamine, 2bl4f1dxvn, aniline, 2,3-dichloro-7ci,8ci, 2,3-dichlorophenylamine
PubChem CID 11844
ChEBI CHEBI:46636
IUPAC Name 2,3-dichloroaniline
SMILES C1=CC(=C(C(=C1)Cl)Cl)N

Specifications

Melting Point 20°C to 24°C
Boiling Point 250°C to 252°C
Flash Point 115°C (239°F)
Quantity 100 g
UN Number UN3442
Beilstein 472027
Formula Weight 162.02
Percent Purity 99%
Chemical Name or Material 2,3-Dichloroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.