missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10169981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10169981 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10169981 Supplier Thermo Scientific Acros Supplier No. 356600050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 82417-45-6
CAS Min % 96.0
CAS Max % 100.0
Boiling Point 140.0°C to 142.0°C (2.0mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Molecular Formula C6H3Cl3O2S
Linear Formula Cl(Cl)C6H3SO2Cl
MDL Number MFCD00051844
Quantity 5g
Synonym 2,3-dichlorobenzene-1-sulfonyl chloride, 2,3-dichlorobenzenesulphonyl chloride, 2,3-dichloro-benzenesulfonyl chloride, 2,3-dichlorobenzenesulphonylchloride, 2,3-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 2,3-dichloro-9ci, 2,3-dichlorobenzenesulfonylchloride, pubchem5114, buttpark 9357-48, acmc-1bl9r
InChI Key PQODWTNHDKDHIW-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl
IUPAC Name 2,3-dichlorobenzenesulfonyl chloride
Molecular Weight (g/mol) 245.51
PubChem CID 2734222
Formula Weight 245.51
Percent Purity 97%
Chemical Name or Material 2, 3-Dichlorobenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dichlorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.