missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorooctafluorobutane, 97%, Thermo Scientific™

Product Code. 11307316
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307316 1 g 1g
11317316 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307316 Supplier Thermo Scientific Alfa Aesar Supplier No. L09252.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 355-20-4
Molecular Formula C4Cl2F8
Molecular Weight (g/mol) 270.93
MDL Number MFCD00018064
InChI Key LXANZHXWGZWFAC-UHFFFAOYNA-N
PubChem CID 78982
IUPAC Name 2,3-dichloro-1,1,1,2,3,4,4,4-octafluorobutane
SMILES FC(F)(F)C(F)(Cl)C(F)(Cl)C(F)(F)F

Specifica

Melting Point ∼-68°C
Density 1.68
Boiling Point 62°C to 63°C
Refractive Index 1.313
Quantity 1 g
Beilstein 1777520
Synonym 2,3-dichlorooctafluorobutane, butane, 2,3-dichloro-1,1,1,2,3,4,4,4-octafluoro, butane, 2,3-dichloro-1,1,1,2,3,4,4,4-octafluoro-, r*,r*, acmc-1cpcx, acmc-20dh57, 2,3-dichlorooctafluorobutan, butane,2,3-dichloro-1,1,1,2,3,4,4,4-octafluoro-, 2r,3r-rel, #, butane,2,3-dichloro-1,1,1,2,3,4,4,4-octafluoro
Formula Weight 270.94
Percent Purity 97%
Chemical Name or Material 2,3-Dichlorooctafluorobutane
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.