missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenyl isothiocyanate, 97%

Product Code. 11469976
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469976 25 g 25g
11459976 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469976 Supplier Thermo Scientific Alfa Aesar Supplier No. B20930.14
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6590-97-2 | C7H3Cl2NS | 204.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6590-97-2
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00041048
InChI Key RQZIODPVCCTBAQ-UHFFFAOYSA-N
PubChem CID 2796281
IUPAC Name 1,2-dichloro-3-isothiocyanatobenzene
SMILES ClC1=CC=CC(N=C=S)=C1Cl

Specifications

Density 1.434
Boiling Point 256°C to 258°C
Flash Point >110°C (230°F)
Odor Irritating
Refractive Index 1.674
Quantity 25 g
UN Number UN2922
Beilstein 2091585
Sensitivity Moisture sensitive
Synonym 2,3-dichlorophenyl isothiocyanate, 2,3-dichlorophenylisothiocyanate, benzene,1,2-dichloro-3-isothiocyanato, benzene, 1,2-dichloro-3-isothiocyanato, dichlorophenylisothiocyanate, dichlorophenyl isothiocyanate, acmc-1b4g1, 2,3-dichlorophenyl-isothiocyanate, 1,2-dichloro-3-isothiocyanato-benzene, 2,3-dichloro-1-isothiocyanato-benzene
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material 2,3-Dichlorophenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.