missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenylacetic acid, 97%, Thermo Scientific™

Product Code. 10570944 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10570944 25 g Glass bottle 25g
10334452 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10570944 Supplier Thermo Scientific Acros Supplier No. 365550250
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10236-60-9
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.04
MDL Number MFCD01861393
InChI Key YWMXEUIQZOQESD-UHFFFAOYSA-N
Synonym 2,3-dichlorophenylacetic acid, 2-2,3-dichlorophenyl acetic acid, benzeneacetic acid, 2,3-dichloro, 2,3-dichlorophenyl acetic acid, 2,3-dichlorobenzeneacetic acid, rarechem al bo 1228, timtec-bb sbb003500, 3-dichlorophenylacetic acid, acmc-1c25x, 2,3-dichlorophenylaceticacid
PubChem CID 2734600
IUPAC Name 2-(2,3-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)Cl)Cl)CC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 130°C to 135°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula Cl2C6H3CH2COOH
Quantity 25 g
Formula Weight 205.04
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 3-Dichlorophenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dichlorophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.