missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloropyridine, 99%

Product Code. 10395980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10395980 50 g Glass bottle 50g
10152570 2.5 g Glass bottle 2.5g
10224380 10 g Glass bottle 10g
11926291 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10395980 Supplier Thermo Scientific Supplier No. 113720500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2402-77-9 | C5H3Cl2N | 147.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2402-77-9
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006229
InChI Key MAKFMOSBBNKPMS-UHFFFAOYSA-N
Synonym 2,3-dichloro-pyridine, pyridine, 2,3-dichloro, dichloropyridine, pyridine, dichloro, ccris 1726, 2,3-dichloro pyridine, pubchem1198, pyridine,3-dichloro, 2.3-dichloro-pyridine, acmc-209g7x
PubChem CID 16988
IUPAC Name 2,3-dichloropyridine
SMILES ClC1=CC=CN=C1Cl

Specifications

Melting Point 67.0°C to 70.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)Cl2
Quantity 50 g
Beilstein 20,II,152
Solubility Information Solubility in water: slightly soluble
Formula Weight 147.99
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Dichloropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dichloropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.