missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloroquinoxaline, 97%

Product Code. 10646501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10646501 10 g Glass bottle 10g
10756642 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10646501 Supplier Thermo Scientific Acros Supplier No. 330830100
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2213-63-0 | C8H4Cl2N2 | 199.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2213-63-0
Molecular Formula C8H4Cl2N2
Molecular Weight (g/mol) 199.04
MDL Number MFCD00006720
InChI Key SPSSDDOTEZKOOV-UHFFFAOYSA-N
Synonym quinoxaline, 2,3-dichloro, 2,3-dichloro-quinoxaline, ccris 4789, quinoxaline, dichloro, dichloroquinoxaline, pubchem15473, 2,3 dichloroquinoxaline, 2.3-dichloroquinoxaline, acmc-1cfck, 3,4-dichloroquinoxaline
PubChem CID 16659
IUPAC Name 2,3-dichloroquinoxaline
SMILES C1=CC=C2C(=C1)N=C(C(=N2)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 150°C to 155°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 10 g
Beilstein 23, 177
Formula Weight 199.04
Percent Purity 97%
Physical Form Needles
Chemical Name or Material 2, 3-Dichloroquinoxaline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dichloroquinoxaline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.