missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dideoxycytidine, 98%

Product Code. 13428668 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13428668 100 mg Glass bottle 100mg
13438668 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13428668 Supplier Thermo Scientific Acros Supplier No. 449501000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7481-89-2 | C9H13N3O3 | 211.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7481-89-2
Molecular Formula C9H13N3O3
Molecular Weight (g/mol) 211.22
MDL Number MFCD00012188
InChI Key WREGKURFCTUGRC-KGQMAECUNA-N
Synonym zalcitabine, 2',3'-dideoxycytidine, dideoxycytidine, ddcyd, hivid, ddc, cytidine, 2',3'-dideoxy, zalcitibine, ddc antiviral, 4-amino-1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one
PubChem CID 24066
ChEBI CHEBI:10101
IUPAC Name 4-amino-1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@H]1CC[C@@H](CO)O1

Specifications

Melting Point 216.0°C to 220.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 1510307
Solubility Information Solubility in water: soluble
Formula Weight 211.22
Percent Purity 98%
Chemical Name or Material 2', 3'-Dideoxycytidine
compliance-icons
Product Identifier
  • 2',3'-Dideoxycytidine
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.