missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dideoxyguanosine, 90+%, Thermo Scientific™

Product Code. p-7043154
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 15437089

Brand: Thermo Scientific Alfa Aesar J65810.MB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85326-06-3
Molecular Formula C10H13N5O3
Molecular Weight (g/mol) 251.25
MDL Number MFCD00057074
InChI Key OCLZPNCLRLDXJC-NTSWFWBYSA-N
Synonym 2',3'-dideoxyguanosine, guanosine, 2',3'-dideoxy, 2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, dideoxyguanosine, 2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6-one, deoxy-deoxy-guanosine, 2,3-didioxyguanosine, 2', 3'-dideoxyguanosine, 2,3-dideoxyguanosine, chembl78101
PubChem CID 64974
IUPAC Name 2-amino-9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES NC1=NC2=C(N=CN2[C@H]2CC[C@@H](CO)O2)C(=O)N1

Specifications

Color White
Quantity 25 mg
Formula Weight 251.24
Percent Purity ≥90%
Physical Form Powder
Chemical Name or Material 2',3'-Dideoxyguanosine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.