missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-methoxybenzaldehyde, 97%, Thermo Scientific™

Catalog Number 11415970
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
256417-11-5
C8H6F2O2
172.13
MFCD04115981
UBBBLLSKHCENQZ-UHFFFAOYSA-N
3744990
2,3-difluoro-4-methoxybenzaldehyde
COC1=C(F)C(F)=C(C=O)C=C1
This item is not returnable. View return policy
256417-11-5
C8H6F2O2
172.13
MFCD04115981
UBBBLLSKHCENQZ-UHFFFAOYSA-N
3744990
2,3-difluoro-4-methoxybenzaldehyde
COC1=C(F)C(F)=C(C=O)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Sensitivity Air Sensitive
Synonym 2,3-difluoro-4-methoxy-benzaldehyde, benzaldehyde, 2,3-difluoro-4-methoxy, acmc-1cri8, benzaldehyde,2,3-difluoro-4-methoxy, 2,3-bis fluoranyl-4-methoxy-benzaldehyde
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material 2,3-Difluoro-4-methoxybenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'