missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 11996481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11996481 1 g 1g
11906491 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11996481 Supplier Thermo Scientific Acros Supplier No. 446780010
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1215034-77-7
Molecular Formula C6H2F5N
Molecular Weight (g/mol) 183.07
InChI Key ZOMVJIOHXSXCIP-UHFFFAOYSA-N
Synonym 2,3-difluoro-4-trifluoromethyl pyridine, alpha,alpha,alpha,2,3-pentafluoro-4-picoline
PubChem CID 13491638
IUPAC Name 2,3-difluoro-4-(trifluoromethyl)pyridine
SMILES C1=CN=C(C(=C1C(F)(F)F)F)F

Specifications

Boiling Point 82.0°C to 84.0°C (400.0 Torr)
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 183.07
Percent Purity 97%
Chemical Name or Material 2, 3-Difluoro-4-(trifluoromethyl)pyridine
compliance-icons compliance-icons
Product Identifier
  • 2,3-Difluoro-4-(trifluoromethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.