missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1,4-benzodioxin-6-ylmethanol, Thermo Scientific™

Product Code. 10262582
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10262582 250 mg 250mg
10150582 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10262582 Supplier Thermo Scientific Maybridge Supplier No. CC01309CB
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 39270-39-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD01317581
InChI Key FFLHNBGNAWYMRH-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzo b 1,4 dioxin-6-yl methanol, 2,3-dihydro-1,4-benzodioxin-6-methanol, 1,4-benzodioxin-6-methanol, 2,3-dihydro, 1,4-benzodioxin-6-methanol,2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-6-yl methanol, 1,4-benzodioxan-6-methanol, 2,3-dihydro-1,4-benzodioxin-6-yl-methanol, 2,3-dihydro-benzo 1,4 dioxin-6-yl methanol, 2,3-dihydrobenzo 1,4 dioxin-6-yl-methanol, 2,3-dihydro-benzo 1,4 dioxin-6-yl-methanol
PubChem CID 2776174
SMILES OCC1=CC=C2OCCOC2=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
IUPAC Name 2,3-dihydro-1,4-benzodioxin-6-ylmethanol
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxin-6-ylmethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.