missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1,4-benzodioxine-5-carboxamide, 97%, Thermo Scientific™

Product Code. 10541112
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10541112 250 mg 250mg
10436282 1 g 1g
10419664 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10541112 Supplier Thermo Scientific Maybridge Supplier No. MO00789CB
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 349550-81-8
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD03659722
InChI Key OIYLTYTXMKZKCR-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzo b 1,4 dioxine-5-carboxamide, maybridge3_004369, 2h,3h-benzo e 1,4-dioxane-5-carboxamide, 1,4-benzodioxin-5-carboxamide,2,3-dihydro, 5wy
PubChem CID 2779845
IUPAC Name 2,3-dihydro-1,4-benzodioxine-5-carboxamide
SMILES C1COC2=C(C=CC=C2O1)C(=O)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 179.17
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-5-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.