missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10171062
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10171062 250 mg 250mg
10762853 1 g 1g
10056654 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10171062 Supplier Thermo Scientific Maybridge Supplier No. CC01301CB
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 4442-54-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00463509
InChI Key JWZQJTGQFHIRFQ-UHFFFAOYSA-N
Synonym 1,4-benzodioxane-6-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-6-carboxylic acid, 3,4-ethylenedioxy benzoic acid, 2,3-dihydro-benzo 1,4 dioxine-6-carboxylic acid, 1,4-benzodioxin-6-carboxylic acid, 2,3-dihydro, 3,4-ethylenedioxybenzoic acid, 1,4-benzodioxan-6-carboxylic acid, 2,3-dihydro-1,4-benzodioxin-6-carboxylic acid, 2,3-dihydro-1,4-benzodioxine-6-carboxylicacid, 6-carboxy-2,3-dihydro-1,4-benzodioxine
PubChem CID 2758833
IUPAC Name 2,3-dihydro-1,4-benzodioxine-6-carboxylic acid
SMILES C1COC2=C(O1)C=CC(=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 180.16
Percent Purity 95%
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-6-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.