missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1-benzofuran-5-yl isocyanate, 97%, Maybridge™

Product Code. 10595825
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10595825 5 g 5g
10212352 250 mg 250mg
10160582 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10595825 Supplier Thermo Scientific Maybridge Supplier No. CC00806DE
 Restricted Item
Estimated Shipment: 19-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 215162-92-8
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD02677718
InChI Key WAIKTAFMGLVRJC-UHFFFAOYSA-N
Synonym 2,3-dihydro-1-benzofuran-5-yl isocyanate, 5-isocyanato-2,3-dihydrobenzofuran, 2,3-dihydro-1-benzofuran-5-ylisocyanate, 2,3-dihydrobenzo b furan-5-isocyanate, benzofuran,2,3-dihydro-5-isocyanato, 2,3-dihydrobenzofuran-5-yl isocyanate, 2,3-dihydro-5-isocyanatobenzo b furan, 5-isocyanato-2,3-dihydrobenzo b furan, 2,3-dihydro-5-isocyanato-1-benzofuran, 2,3-dihydro-1-benzofuran-5-yl-isocyanate
PubChem CID 2776156
IUPAC Name 5-isocyanato-2,3-dihydro-1-benzofuran
SMILES C1COC2=C1C=C(C=C2)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 161.16
Percent Purity 95%
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-5-yl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.