missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1-benzofuran-5-ylboronic acid, 95%, May contain varying amounts of anhydride, Thermo Scientific™

Product Code. 10754692
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10754692 5 g 5g
10190772 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10754692 Supplier Thermo Scientific Maybridge Supplier No. CC00812DE
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 227305-69-3
Molecular Formula C8H9BO3
Molecular Weight (g/mol) 163.97
MDL Number MFCD02681979
InChI Key ZIXLJHSFAMVHPC-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-boronic acid, 2,3-dihydrobenzofuran-5-ylboronic acid, 2,3-dihydrobenzofuran-5-yl boronic acid, 2,3-dihydrobenzo b furan-5-boronic acid, 2,3-dihydrobenzofuran-5-yl-5-boronic acid, boronic acid, 2,3-dihydro-5-benzofuranyl, 2,3-dihydro-1-benzofuran-5-yl-boranediol, 2,3-dihydro-1-benzofuran-5-yl boronic acid, pubchem7848, zlchem 1108
PubChem CID 2773380
SMILES OB(O)C1=CC=C2OCCC2=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 5 g
IUPAC Name (2,3-dihydro-1-benzofuran-5-yl)boronic acid
Formula Weight 163.97
Percent Purity 95%
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-5-ylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.