missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-3-hydroxy-2-oxo Lysergide, TRC

Product Code. 18090050
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18090050 10 mg 10mg
18021030 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18090050 Supplier Toronto Research Chemicals Supplier No. TRCD44956010MG
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111295-09-1
Molecular Formula C20 H25 N3 O3
Molecular Weight (g/mol) 355.43
Synonym 2-Oxo-3-hydroxy-LSD, 2,3-Dihydro-3-hydroxy-2-oxo Lysergide
IUPAC Name (6aR,9R)-N,N-diethyl-5a-hydroxy-7-methyl-5-oxo-6,6a,8,9-tetrahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide
SMILES CCN(CC)C(=O)[C@H]1CN(C)[C@@H]2CC3(O)C(=O)Nc4cccc(C2=C1)c34

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H25N3O3/c1-4-23(5-2)18(24)12-9-14-13-7-6-8-15-17(13)20(26,19(25)21-15)10-16(14)22(3)11-12/h6-9,12,16,26H,4-5,10-11H2,1-3H3,(H,21,25)/t12-,16-,20?/m1/s1
Formula Weight 355.1896
Chemical Name or Material 2-Oxo-3-hydroxy-LSD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.