missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzo[b]furan-5-carbothioamide, 97%, Thermo Scientific™

Product Code. 10783137
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10783137 5 g 5g
10233382 250 mg 250mg
10039601 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10783137 Supplier Thermo Scientific Maybridge Supplier No. SP00108DE
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 306936-08-3
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.237
MDL Number MFCD00728868
InChI Key SOAROQIQNPHLJX-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzo b furan-5-carbothioamide, 2,3-dihydrobenzofuran-5-carbothioamide, 2,3-dihydrobenzo b furan-5-thioamide, maybridge1_008543, 2,3-dihydrobenzofuran-5-thioamide, 2,3-dihydro-1-benzofuran-5-thiocarboxamide, 2,3-dihydrobenzo b furan-5-carbothioic acid amide, amino-2,3-dihydrobenzo b furan-5-ylmethane-1-thione
PubChem CID 2736118
IUPAC Name 2,3-dihydro-1-benzofuran-5-carbothioamide
SMILES C1COC2=C1C=C(C=C2)C(=S)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 179.24
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-5-carbothioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.