missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzo[b]furan-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10516314
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10516314 10 g Amber Glass Bottle 10g
10642465 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10516314 Supplier Thermo Scientific Maybridge Supplier No. CC00801EA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 76429-73-7
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
InChI Key YXYOLVAXVPOIMA-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-carboxylic acid, 2,3-dihydrobenzo b furan-5-carboxylic acid, 5-benzofurancarboxylic acid, 2,3-dihydro, pubchem9983, pubchem14824, maybridge1_002497, ksc377a5f, dihydrobenzofuran-5-carboxylic acid, 5-carboxy-2,3-dihydrobenzo b furan
PubChem CID 676583
IUPAC Name 2,3-dihydro-1-benzofuran-5-carboxylic acid
SMILES C1COC2=C1C=C(C=C2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.