missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzo[b]furan-7-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10659174
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10659174 250 mg Amber Glass Bottle 250mg
10236182 1 g Amber Glass Bottle 1g
10633795 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10659174 Supplier Thermo Scientific Maybridge Supplier No. CC00901CB
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 35700-40-4
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00671526
InChI Key UHXBMSNEECJPSX-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-7-carboxylic acid, 2,3-dihydrobenzo b furan-7-carboxylic acid, 7-coumarancarboxylic acid, 2,3-dihydro-7-benzofurancarboxylic acid, 7-benzofurancarboxylicacid, 2,3-dihydro, 7-benzofurancarboxylic acid, 2,3-dihydro, pubchem16734, acmc-209iiq, dihydrobenzofurancarboxylicacid, 7-dihydrobenzofuran carboxylic acid
PubChem CID 2795014
IUPAC Name 2,3-dihydro-1-benzofuran-7-carboxylic acid
SMILES C1COC2=C(C=CC=C21)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-7-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.