missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde, ≥97%, Thermo Scientific™

Product Code. 10187892
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10187892 250 mg 250mg
10012134 1 g 1g
10215534 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10187892 Supplier Thermo Scientific Maybridge Supplier No. CC47804CB
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 204905-77-1
Molecular Formula C7H6O3S
Molecular Weight (g/mol) 170.182
MDL Number MFCD01651766
InChI Key GNVXYRDVJKJZTO-UHFFFAOYSA-N
Synonym 2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carbaldehyde, 2h,3h-thieno 3,4-b 1,4 dioxine-5-carbaldehyde, 3,4-ethylenedioxythiophene-2-carbaldehyde, 3,4-ethylenedioxythiophene-2-carboxaldehyde, 2-formyl-3,4-ethylene dioxythiophene, 2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carbaldehyde, 2,3-dihydrothieno 3,4-b-1,4-dioxin-5-carbaldehyde, 2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxaldehyde, thieno 3,4-b-1,4-dioxin-5-carboxaldehyde, 2,3-dihydro
PubChem CID 3540090
IUPAC Name 2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde
SMILES C1COC2=C(SC=C2O1)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 170.19
Percent Purity 97%
Chemical Name or Material 2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.