missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxybenzaldehyde, 98+%

Product Code. 11458016
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458016 100 g 100g
11448016 25 g 25g
11468016 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458016 Supplier Thermo Scientific Alfa Aesar Supplier No. A14080.22
 Restricted Item
Estimated Shipment: 07-12-2026
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 86-51-1 | C9H10O3 | 166.176 g/mol

2,3-Dimethoxybenzaldehyde possess antifungal activity of redox-active benzaldehydes that target cellular antioxidation. It could function as chemosensitizing agents in concert with conventional drugs to improve antifungal efficacy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethoxybenzaldehyde possess antifungal activity of redox-active benzaldehydes that target cellular antioxidation. It could function as chemosensitizing agents in concert with conventional drugs to improve antifungal efficacy.

Solubility
Solubility in methanol with almost transparency.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 86-51-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00003309
InChI Key JIVGSHFYXPRRSZ-UHFFFAOYSA-N
Synonym o-veratraldehyde, benzaldehyde, 2,3-dimethoxy, unii-8alp3sy00l, 2,3-dimethoxy-benzaldehyde, 8alp3sy00l, benzaldehyde,3-dimethoxy, dimethoxybenzaldehyde, pubchem8216, benzaldehyde, dimethoxy, upcmld00wstructure66
PubChem CID 66581
IUPAC Name 2,3-dimethoxybenzaldehyde
SMILES COC1=CC=CC(=C1OC)C=O

Specifikationer

Melting Point 51°C to 52°C
Boiling Point 178°C (20 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 908264
Sensitivity Air Sensitive
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 2,3-Dimethoxybenzaldehyde

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.