missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-1,3-butadiene, 98%, stabilized with BHT

Produktkod. 10239653 Handla allt Thermo Scientific Chemicals Produkter
Change view
Klicka för att se tillgängliga alternativ
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Förpackningsstorlek:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Packaging unitSize
10239653 5 g Glass bottle 5g
10065823 25 g Glass bottle 25g
10025873 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10239653 Leverantör Thermo Scientific Acros Leverantörsnummer 163430050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 513-81-5 | C6H10 | 82.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 513-81-5
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00008595
InChI Key SDJHPPZKZZWAKF-UHFFFAOYSA-N
Synonym 2,3-dimethyl-1,3-butadiene, biisopropenyl, 2,3-dimethylbutadiene, diisopropenyl, 2,3-dimethylenebutane, 1,3-butadiene, 2,3-dimethyl, unii-61tuu25hco, 61tuu25hco, 2,3-dimethyl-buta-1,3-diene, 2,3-diene
PubChem CID 10566
IUPAC Name 2,3-dimethylbuta-1,3-diene
SMILES CC(=C)C(C)=C

Specifikationer

Melting Point -76.0°C
Color Colorless
Density 0.7260g/mL
Boiling Point 68.0°C to 69.0°C
Flash Point −20°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)C(CH3)=CH2
Refractive Index 1.438
Quantity 5 g
Beilstein 01, III, 991
Fieser 01,276; 13,45
Merck Index 15, 3260
Specific Gravity 0.726
Stabilizer 100ppm BHT
Formula Weight 82.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 3-Dimethyl-1, 3-butadiene, Stabilized with BHT
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • 2,3-Dimethyl-1,3-butadiene
Signalord
  • Danger
Riskkategorier
  • Flammable liquid Category 2
Faroangivelser
  • H225-Highly flammable liquid and vapour.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Kompletterande information
  • MIXTURE LIST-Contain: 2,3-dimethyl-1,3-Butadiene

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.