missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-1,3-butadiene, TRC

Product Code. 30381483
Change view
Click to view available options
Quantity:
1 g
missing translation for 'unitSize'
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381483 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30381483 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCD4785351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dimethyl-1,3-butadiene
CAS 513-81-5
Molecular Formula C6H10
Quantity 1 g
InChI Formula InChI=1S/C6H10/c1-5(2)6(3)4/h1,3H2,2,4H3
SMILES CC(=C)C(=C)C
IUPAC Name 2,3-dimethylbuta-1,3-diene
Molecular Weight (g/mol) 82.14
Formula Weight 82.0783
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.