missing translation for 'onlineSavingsMsg'
Learn More

2,3-dimethyl-1h-indole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10738825
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10738825 250 mg 250mg
10182974 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10738825 Supplier Thermo Scientific Maybridge Supplier No. MO07746CB
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 14844-73-6
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.214
InChI Key KHJGIMZYCBPOBG-UHFFFAOYSA-N
PubChem CID 693386
IUPAC Name 2,3-dimethyl-1H-indole-5-carboxylic acid
SMILES CC1=C(NC2=C1C=C(C=C2)C(=O)O)C

Specifications

Assay Percent Range 97%
Quantity 250 mg
Synonym 1h-indole-5-carboxylic acid, 2,3-dimethyl, 2,3-dimethylindole-5-carboxylic acid, 5-carboxy-2,3-dimethyl-1h-indole, 2,3-dimethyl-indole-5-carboxylic acid, 1h-indole-5-carboxylicacid,2,3-dimethyl, 1h-indole-5-carboxylicacid, 2,3-dimethyl, 2,3-dimethyl-1h-indole-5-carboxylic acid
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material 2, 3-Dimethyl-1H-indole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.