missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-1H-indole-6-carboxylic acid, 97%, Thermo Scientific™

Codice prodotto. 10786903
Cambia vista
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Dimensione della confezione:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Packaging Dimensione della confezione
10786903 1 g Amber Glass Bottle 1g
10192784 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10786903 Fornitore Thermo Scientific Maybridge N. del fornitore MO07747DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Identificatori chimici

CAS 103986-06-7
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.214
MDL Number MFCD06799503
InChI Key UDPOIMGOHQYJEE-UHFFFAOYSA-N
Synonym 1h-indole-6-carboxylicacid, 2,3-dimethyl, 2,3-dimethylindole-6-carboxylic acid, acmc-1bpsb, 2,3-dimethyl-6-carboxyindole, 1h-indole-6-carboxylic acid, 2,3-dimethyl
PubChem CID 2772885
IUPAC Name 2,3-dimethyl-1H-indole-6-carboxylic acid
SMILES CC1=C(NC2=C1C=CC(=C2)C(=O)O)C

Specifica

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material 2, 3-Dimethyl-1H-indole-6-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.