missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethyl-2-butene, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Pakningsstørrelse:
100mL
25mL
500mL
Beskrivelse
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | -75.0°C |
| Color | Colorless |
| Density | 0.7000g/mL |
| Boiling Point | 73.0°C |
| Flash Point | −16°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- 2,3-Dimethyl-2-butene
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion