missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-2-butene, TRC

Product Code. 30380245
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Product Code. Quantity unitSize
30380245 50 mL 50mL
1 options
This item is not returnable. View return policy

Product Code. 30380245

Brand: Toronto Research Chemicals TRCD47197550ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dimethyl-2-butene
CAS 563-79-1
Molecular Formula C6H12
Quantity 50 mL
Synonym 1,1,2,2-Tetramethylethylene, NSC 73907, Tetramethylethene, Tetramethylethylene
InChI Formula InChI=1S/C6H12/c1-5(2)6(3)4/h1-4H3
SMILES CC(=C(C)C)C
IUPAC Name 2,3-dimethylbut-2-ene
Molecular Weight (g/mol) 84.16
Formula Weight 84.0939
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.