missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylaniline, 98%

Product Code. 11484303
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484303 2500 g 2500g
11474303 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484303 Supplier Thermo Scientific Alfa Aesar Supplier No. A10918.0E
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 87-59-2 | C8H11N | 121.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-59-2
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00007732
InChI Key VVAKEQGKZNKUSU-UHFFFAOYSA-N
Synonym 2,3-xylidine, 3-amino-o-xylene, benzenamine, 2,3-dimethyl, 2,3-dimethylbenzenamine, 2,3-dimethylphenylamine, 2,3-xylylamine, ortho-xylidine, vic-o-xylidine, unii-zd450abi9x, ccris 4739
PubChem CID 6893
IUPAC Name 2,3-dimethylaniline
SMILES CC1=C(C(=CC=C1)N)C

Specifications

Melting Point 2°C to 3°C
Density 0.992
Quantity 2500 g
Odor Characteristic
Refractive Index 1.568
UN Number UN1711
Beilstein 742174
Merck Index 14,10084
Boiling Point 221°C to 222°C
Formula Weight 121.18
Percent Purity 98%
Chemical Name or Material 2,3-Dimethylaniline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,3-Dimethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P308-IF exposed or concerned:.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.