missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylaniline, 98%

Product Code. p-7081099
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
This item is not returnable. View return policy

Product Code. 11474303

Brand: Thermo Scientific Alfa Aesar A10918.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87-59-2 | C8H11N | 121.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-59-2
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00007732
InChI Key VVAKEQGKZNKUSU-UHFFFAOYSA-N
Synonym 2,3-xylidine, 3-amino-o-xylene, benzenamine, 2,3-dimethyl, 2,3-dimethylbenzenamine, 2,3-dimethylphenylamine, 2,3-xylylamine, ortho-xylidine, vic-o-xylidine, unii-zd450abi9x, ccris 4739
PubChem CID 6893
IUPAC Name 2,3-dimethylaniline
SMILES CC1=C(C(=CC=C1)N)C

Specifications

Melting Point 2°C to 3°C
Density 0.992
Boiling Point 221°C to 222°C
Quantity 500 g
Odor Characteristic
Refractive Index 1.568
UN Number UN1711
Beilstein 742174
Merck Index 14,10084
Formula Weight 121.18
Percent Purity 98%
Chemical Name or Material 2,3-Dimethylaniline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,3-Dimethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P308-IF exposed or concerned:.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.