missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbenzeneboronic acid, 98%

Product Code. 11426816
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426816 1 g 1g
11436816 5 g 5g
11446816 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426816 Supplier Thermo Scientific Alfa Aesar Supplier No. B23942.03
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 183158-34-1 | C8H11BO2 | 149.984 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 183158-34-1
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.984
MDL Number MFCD01863524
InChI Key ZYYANAWVBDFAHY-UHFFFAOYSA-N
Synonym 2,3-dimethylbenzeneboronic acid, 2,3-dimethylphenyl boronic acid, o-xylene-3-boronic acid, 2,3-dimethylphenyl boranediol, 2,3-dimethylbenzeneboronicacid, 2,3-dimethylphenylboronicacid, pubchem5037, acmc-1bxk2, amtb394, ksc174k9l
PubChem CID 2773395
IUPAC Name (2,3-dimethylphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)C)C)(O)O

Specifications

Melting Point ∼175°C
Quantity 1 g
Beilstein 8050763
Formula Weight 149.99
Percent Purity 98%
Chemical Name or Material 2,3-Dimethylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.