missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbenzoyl chloride, 96%

Product Code. 11475610
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11475610

Brand: Thermo Scientific Alfa Aesar H25873.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 21900-46-9 | C9H9ClO | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21900-46-9
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00045217
InChI Key WFNKMVDATNLZBX-UHFFFAOYSA-N
Synonym 2,3-dimethylbenzene-1-carbonyl chloride, benzoyl chloride, dimethyl, xylic acid chloride, acmc-20aog3, 2,3 dimethylbenzoyl chloride, 2,3-dimethyl-benzoyl chloride, 2,3-dimethylbenzene-1-carbonylchloride, 2,3-dimethyl-1-benzenecarbonyl chloride, benzoyl chloride, 2,3-dimethyl-6ci,7ci,8ci,9ci
PubChem CID 2800899
IUPAC Name 2,3-dimethylbenzoyl chloride
SMILES CC1=CC=CC(=C1C)C(=O)Cl

Specifications

Melting Point 2°C
Boiling Point 122°C to 124°C
Quantity 25 g
Refractive Index 1.555
UN Number UN3265
Formula Weight 168.62
Percent Purity 96%
Chemical Name or Material 2,3-Dimethylbenzoyl chloride

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado