missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbenzoyl chloride, 96%

Product Code. 11485610
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485610 100 g 100g
11465610 5 g 5g
11475610 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485610 Supplier Thermo Scientific Alfa Aesar Supplier No. H25873.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 21900-46-9 | C9H9ClO | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21900-46-9
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00045217
InChI Key WFNKMVDATNLZBX-UHFFFAOYSA-N
Synonym 2,3-dimethylbenzene-1-carbonyl chloride, benzoyl chloride, dimethyl, xylic acid chloride, acmc-20aog3, 2,3 dimethylbenzoyl chloride, 2,3-dimethyl-benzoyl chloride, 2,3-dimethylbenzene-1-carbonylchloride, 2,3-dimethyl-1-benzenecarbonyl chloride, benzoyl chloride, 2,3-dimethyl-6ci,7ci,8ci,9ci
PubChem CID 2800899
IUPAC Name 2,3-dimethylbenzoyl chloride
SMILES CC1=CC=CC(=C1C)C(=O)Cl

Specifications

Melting Point 2°C
Quantity 100 g
Refractive Index 1.555
UN Number UN3265
Boiling Point 122°C to 124°C
Formula Weight 168.62
Percent Purity 96%
Chemical Name or Material 2,3-Dimethylbenzoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.