missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbutane, 98+%

Product Code. 13448318 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13448318 1 L Glass Bottle 1 L
10439410 5 mL Glass Bottle 5mL
10239783 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 13448318 Supplier Thermo Scientific Acros Supplier No. 116100010
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 79-29-8 | C6H14 | 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-29-8
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00008925
InChI Key ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
Synonym diisopropyl, biisopropyl, butane, 2,3-dimethyl, 1,1,2,2-tetramethylethane, hsdb 76, unii-68isq7a432, ccris 6020, ch3 2chch ch3 2, acmc-1bmfq, dsstox_cid_5112
PubChem CID 6589
IUPAC Name 2,3-dimethylbutane
SMILES CC(C)C(C)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -129.0°C
Color Colorless
Density 0.6620g/mL
Boiling Point 58.0°C
Flash Point −29°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Linear Formula (CH3)2CHCH(CH3)2
Refractive Index 1.3740 to 1.3760
Quantity 1 L
UN Number 2457
Beilstein 01,151
Specific Gravity 0.662
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 86.18
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2, 3-Dimethylbutane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,3-Dimethylbutane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.