missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylmaleic anhydride, 97%

Product Code. 11323446
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323446 5 g 5g
11333446 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323446 Supplier Thermo Scientific Alfa Aesar Supplier No. L12919.06
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 766-39-2 | C6H6O3 | 126.111 g/mol

2,3-Dimethylmaleic anhydride acts as a reagent in the preparation of maleimides. It is also employed as an amino group protecting agent for superoxide dismutase. Further, it serves as a reactant in the dissociation of ribosomal proteins. In addition to this, it is used in the preparation of dimethylmaleic acid dimethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethylmaleic anhydride acts as a reagent in the preparation of maleimides. It is also employed as an amino group protecting agent for superoxide dismutase. Further, it serves as a reactant in the dissociation of ribosomal proteins. In addition to this, it is used in the preparation of dimethylmaleic acid dimethyl ester.

Solubility
Soluble in ethanol, ether and chloroform. Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-39-2
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00005523
InChI Key MFGALGYVFGDXIX-UHFFFAOYSA-N
Synonym 2,3-dimethylmaleic anhydride, dimethylmaleic anhydride, 2,5-furandione, 3,4-dimethyl, pyrocinchonic anhydride, maleic anhydride, dimethyl, dimethylmaleic acid anhydride, 2,3-dimethylmaleicanhydride, unii-6pp3n541qa, 3,4-dimethyl-furan-2,5-dione, alpha,beta-dimethylmaleic anhydride
PubChem CID 13010
IUPAC Name 3,4-dimethylfuran-2,5-dione
SMILES CC1=C(C(=O)OC1=O)C

Specifications

Melting Point 93°C to 96°C
Boiling Point 222°C to 223°C
Quantity 5 g
Beilstein 112044
Sensitivity Moisture sensitive
Solubility Information Soluble in ethanol,ether and chloroform. Slightly soluble in water.
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylmaleic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.