missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylnaphthalene, 97%

Product Code. 15545036 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15545036 1 g 1g
15555036 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15545036 Supplier Thermo Scientific Acros Supplier No. 116450010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 581-40-8 | C12H12 | 156.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 581-40-8
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
InChI Key WWGUMAYGTYQSGA-UHFFFAOYSA-N
Synonym guajen, naphthalene, 2,3-dimethyl, unii-35dj6se17o, 2,3-dmn, 2,3-dimethyl naphthalene, acmc-1ao3c, 2,3-dimethylnaphthalene, naphthalene, 2,3-dimethyl-8ci 9ci
PubChem CID 11386
ChEBI CHEBI:48615
IUPAC Name 2,3-dimethylnaphthalene
SMILES CC1=CC2=CC=CC=C2C=C1C

Specifications

Melting Point 102.0°C to 104.0°C
Boiling Point 269.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula C10H6(CH3)2
Quantity 1 g
Beilstein 05,IV,1713
Solubility Information Solubility in water: insoluble.
Formula Weight 156.23
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylnaphthalene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.