missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylnaphthalene, 98%, BLD Pharm™

Product Code. 31102431
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31102431 100 mg
31104446 250 mg
31099380 5 g
31110389 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31102431 Supplier BLD Pharm Supplier No. BD13846100mg
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 581-40-8
Molecular Formula C12H12
MDL Number MFCD00004119
Quantity 100 mg
SMILES CC1=C(C)C=C2C=CC=CC2=C1
Molecular Weight (g/mol) 156.22
Percent Purity 98%
Chemical Name or Material 2,3-Dimethylnaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.