missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylphenyl isocyanate, 99%

Product Code. 11388305
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388305 5 g 5g
11378305 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388305 Supplier Thermo Scientific Alfa Aesar Supplier No. L10225.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1591-99-7 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1591-99-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00013851
InChI Key KNHJIEOCVVIBIV-UHFFFAOYSA-N
Synonym 2,3-dimethylphenyl isocyanate, 2,3-dimethylphenylisocyanate, benzene, isocyanatodimethyl, 2,3-dimethylbenzenisocyanate, dimethylphenyl isocyanate, acmc-1bttc, 1-isocyanato-2,3-dimethyl-benzene, #, benzene,1-isocyanato-2,3-dimethyl
PubChem CID 137096
ChEBI CHEBI:63899
IUPAC Name 1-isocyanato-2,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)N=C=O)C

Specifications

Density 1.059
Quantity 5 g
Odor Pungent
Refractive Index 1.54
UN Number UN2206
Beilstein 6313438
Boiling Point 104°C to 105°C (50 mmHg)
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material 2,3-Dimethylphenyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.