missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylphenyl isocyanate, 99%

Product Code. p-7045309
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11378305

Brand: Thermo Scientific Alfa Aesar L10225.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1591-99-7 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1591-99-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00013851
InChI Key KNHJIEOCVVIBIV-UHFFFAOYSA-N
Synonym 2,3-dimethylphenyl isocyanate, 2,3-dimethylphenylisocyanate, benzene, isocyanatodimethyl, 2,3-dimethylbenzenisocyanate, dimethylphenyl isocyanate, acmc-1bttc, 1-isocyanato-2,3-dimethyl-benzene, #, benzene,1-isocyanato-2,3-dimethyl
PubChem CID 137096
ChEBI CHEBI:63899
IUPAC Name 1-isocyanato-2,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)N=C=O)C

Specifications

Density 1.059
Boiling Point 104°C to 105°C (50 mmHg)
Quantity 1 g
Odor Pungent
Refractive Index 1.54
UN Number UN2206
Beilstein 6313438
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material 2,3-Dimethylphenyl isocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.