missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diphenylmaleic anhydride, 98%

Product Code. 10412750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10412750 1 g 1g
10371510 5 g 5g
10179929 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10412750 Supplier Thermo Scientific Acros Supplier No. 439010010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4808-48-4 | C16H10O3 | 250.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4808-48-4
Molecular Formula C16H10O3
Molecular Weight (g/mol) 250.26
InChI Key OUJCFCNZIUTYBH-UHFFFAOYSA-N
Synonym 2,3-diphenylmaleic anhydride, diphenylmaleic anhydride, 3,4-diphenyl-2,5-furandione, diphenylfuran-2,5-dione, 2,3-diphenylmaleicanhydride, 2,5-furandione, 3,4-diphenyl, acmc-1asck, 2, 3,4-diphenyl, diphenyl-maleic anhydride, 2,5-furandione,3,4-diphenyl
PubChem CID 78530
IUPAC Name 3,4-diphenylfuran-2,5-dione
SMILES C1=CC=C(C=C1)C2=C(C(=O)OC2=O)C3=CC=CC=C3

Spécification

CAS Min % 97.5
CAS Max % 100.0
Melting Point 157.1°C to 159.4°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 250.26
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2, 3-Diphenylmaleic anhydride
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 2,3-Diphenylmaleic anhydride
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.