missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Methoxyphenyl)ethylamine, 97+%

Product Code. 15411739
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15411739 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15411739 Supplier Thermo Scientific Alfa Aesar Supplier No. H64494.22
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2039-67-0 | C9H13NO | 151.209 g/mol

Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2039-67-0
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00008187
InChI Key WJBMRZAHTUFBGE-UHFFFAOYSA-N
Synonym 3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine
PubChem CID 74866
IUPAC Name 2-(3-methoxyphenyl)ethanamine
SMILES COC1=CC=CC(=C1)CCN

Specifications

Color Yellow
Density 1.038 g/mL
Boiling Point 118°C to 118°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.538
Quantity 100 g
UN Number UN2735
Beilstein 775202
Solubility Information Insoluble in water.
Formula Weight 151.21
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 2-(3-Methoxyphenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.