missing translation for 'onlineSavingsMsg'
Learn More

2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridyl]methyl]sulfinyl]-5-methoxy-O- methyl-1H-benzimidazole, TRC

Product Code. 30478241
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478241 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478241 Supplier Toronto Research Chemicals Supplier No. TRCM33091050MG
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[([3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridyl]methyl)sulfinyl]-5-methoxy-O-methyl-1H-benzimidazole
CAS 1184993-29-0
Molecular Formula C18 H18 F3 N3 O4 S
Quantity 50 mg
Synonym 5-Hydroxy Lansoprazole Methoxymethyl Ether
InChI Formula InChI=1S/C18H18F3N3O4S/c1-11-15(22-6-5-16(11)27-9-18(19,20)21)8-29(25)17-23-13-4-3-12(28-10-26-2)7-14(13)24-17/h3-7H,8-10H2,1-2H3,(H,23,24)
SMILES COCOc1ccc2nc([nH]c2c1)S(=O)Cc3nccc(OCC(F)(F)F)c3C
IUPAC Name 6-(methoxymethoxy)-2-[[3-methyl-4-(2,2,2-trifluoroethoxy)-2-pyridyl]methylsulfinyl]-1H-benzimidazole
Molecular Weight (g/mol) 429.41
Formula Weight 429.097
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.