missing translation for 'onlineSavingsMsg'
Learn More

2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole, 98%

Product Code. 15435535
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435535 1 g 1g
15455535 250 mg 250mg
15445535 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15435535 Supplier Thermo Scientific Alfa Aesar Supplier No. H61878.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103577-40-8 | C16H14F3N3OS | 353.363 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103577-40-8
Molecular Formula C16H14F3N3OS
Molecular Weight (g/mol) 353.363
MDL Number MFCD00834357
InChI Key CCHLMSUZHFPSFC-UHFFFAOYSA-N
Synonym lansoprazole sulfide, 2-3-methyl-4-2,2,2-trifluoroethoxy pyridin-2-yl methyl thio-1h-benzo d imidazole, 2-3-methyl-4-2,2,2-trifluoroethoxy-2-pyridinyl methylthio-1h-benzimidazole, unii-58xp950e2t, lansoprazole sulphide compound, 2-3-methyl-4-2,2,2-trifluoroethoxy-2-pyridinyl methyl thio-1h-benzimidazole, 2-methyl-4-2,2,2-trifluoroethoxy pyridinyl thio-1h-binzimidazole, 1h-benzimidazole,2-3-methyl-4-2,2,2-trifluoroethoxy-2-pyridinyl methyl thio, 2-3-methyl-4-2,2,2-trifluoroethoxy pyridin-2-yl methyl sulfanyl-1h-1,3-benzodiazole, 2-3-methyl-4-2,2,2-trifluoroethoxy pyridinyl methyl thio-1h-benzimidazole
PubChem CID 1094080
IUPAC Name 2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfanyl]-1H-benzimidazole
SMILES CC1=C(C=CN=C1CSC2=NC3=CC=CC=C3N2)OCC(F)(F)F

Specifications

Quantity 1 g
Formula Weight 353.36
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.