missing translation for 'onlineSavingsMsg'
Learn More

2,3-O-Isopropylidene-D-erythronolactone, 98%, Thermo Scientific Chemicals

Product Code. 15498399
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498399 5 g 5g
15488399 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498399 Supplier Thermo Scientific Alfa Aesar Supplier No. J66871.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

For use in spiroannulated carbohydrate synthesis

It undergoes Aldol condensations with silyl ketene acetals.1 Employed in spiroannulated carbohydrate synthesis.2 Convergent syntheses of a hydroxylated indolizidine,3 carbohydrate substituted benzoquinones,4 and of the oxazole segment of calyculin5 have been accomplished using this chiral synthon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It undergoes Aldol condensations with silyl ketene acetals.1 Employed in spiroannulated carbohydrate synthesis.2 Convergent syntheses of a hydroxylated indolizidine,3 carbohydrate substituted benzoquinones,4 and of the oxazole segment of calyculin5 have been accomplished using this chiral synthon.

Solubility
Soluble in water

Notes
Store at room temperature. Incompatible with heat, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25581-41-3
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.15
MDL Number MFCD00134440
InChI Key WHPSMBYLYRPVGU-UHFFFAOYNA-N
Synonym 2,3-o-isopropylidene-d-erythronolactone, --2,3-o-isopropylidene-d-erythronolactone, 3ar,6ar-2,2-dimethyldihydrofuro 3,4-d 1,3 dioxol-4 3ah-one, 3ar,6ar-2,2-dimethyl-6,6a-dihydro-3ah-furo 3,4-d 1,3 dioxol-4-one, 3ar,6ar-2,2-dimethyl-dihydro-3ah-furo 3,4-d 1,3 dioxol-4-one, pubchem19725, 2,3-o-isopropylidene-d-erythronolactol, d-2,3-o-isopropylidene-erythronic acid, a-lactone, 2-o,3-o-isopropylidene-d-erythronic acid gamma-lactone
PubChem CID 386843
IUPAC Name (3aR,6aR)-2,2-dimethyl-6,6a-dihydro-3aH-furo[3,4-d][1,3]dioxol-4-one
SMILES CC1(C)OC2COC(=O)C2O1

Specifications

Melting Point 67°C to 69°C
Color White
Quantity 5 g
Beilstein 1282952
Solubility Information Soluble in water
Optical Rotation −118° (c=1 in Water)
Formula Weight 158.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3-O-Isopropylidene-D-erythronolactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.